Attention:
Certain features of sigmaaldrich.com will be down for maintenance Saturday afternoon/evening, February 24th starting at 3:00 pm CT until 9:00 pm CT.
Please note that you still have telephone and email access to our local offices. We apologize for any inconvenience.
Discover Unprotected Amino Aldehydes from Professor Yudin
By: Mark Redlich, Chemfiles Volume 9 Article 3
Professor Andrei Yudin and co-workers have recently described the preparation of bench-stable, unprotected α-amino aldehydes.1These kinetically amphoteric molecules exist as dimers, and due to the strain of the aziridine ring, resist inter- and intramolecular iminium ion formation. Furthermore, the two functionalities remain orthogonal to each other throughout their transformations, allowing for the reaction of the aldehyde without the requirement of an additional protecting group.
α-amino aldehydes
Whereas the reductive amination of protected amino aldehydes has significant limitations due to epimerization or overalkylation, these Yudin amino aldehyde dimers do not suffer from either limitation, due to a negligible concentration of free aldehyde during the reaction. This allows the researcher facile access to a method for the creation of complex polycyclic skeletons2 or peptidomimetic conjugates3 with a high degree of stereocontrol. Nucleophilic additions,4Wittig and related olefination reactions can be carried out with high selectivities and yields. Sigma-Aldrich is pleased to offer these useful Yudin amino aldehydes for your research.
Cascade Polycyclizations
Cascade Polycyclizations
Synthesis of Peptidomimetic Conjugates Without Protecting Groups
Synthesis of Peptidomimetic Conjugates Without Protecting Groups
Unprotected Vinyl Aziridines via Olefination
Unprotected Vinyl Aziridines via Olefination
Unprotected Vicinal 1,2-Amino Alcohols via Allylation with Indium Reagents
Unprotected Vicinal 1,2-Amino Alcohols via Allylation with Indium Reagents
back to top 
Materials
References
  1. Hili, R.; Yudin, A. K. J. Am. Chem. Soc. 2006, 128, 14772.
  2. Yudin, A. K.; Hili, R. Chem.—Eur. J. 2007, 13, 6538.
  3. Li, X.; Yudin, A. K. J. Am. Chem. Soc. 2007, 129, 14152.
  4. Hili, R.; Yudin, A. K. Angew. Chem., Int. Ed. 2008, 47, 4188.