Ethyl 5-[(4-methylphenyl)sulfonyl]-3-oxopentanoate: A Bench-Stable Precursor for Nazarov's Reagent
By: Dr. Mark Redlich, Chemfiles Volume 10 Article 1
Nazarov's reagent is a commonly used annulating agent, but its synthesis is often fraught with poor yields or difficulties isolating the material. De Risi and coworkers recently reported the synthesis and application of ethyl 5-[(4-methylphenyl)sulfonyl]- 3-oxopentanoate, which is a bench-stable powder that can be stored at room temperature without any special precautions, and demasked under various conditions to form ethyl 3-oxopent- 4-enoate (Nazarov's reagent) in situ to generate annulated products (Scheme 6).1
Scheme 6
Scheme 6: (718327)
back to top
  1. Benetti, S. et al. Synlett 2008, 2609.