Attention:
Certain features of sigmaaldrich.com will be down for maintenance Saturday afternoon/evening, February 24th starting at 3:00 pm CT until 9:00 pm CT.
Please note that you still have telephone and email access to our local offices. We apologize for any inconvenience.
Ferrocenyl Based Ligands and Catalysts
By: Josephine Nakhla, Aldrich ChemFiles 2009, 9.2, 11.
Aldrich ChemFiles 2009, 9.2, 11.
Hartwig and co-workers have reported the use of the electron-rich and bulky ligand 1,1’-bis(di-tert-butylphosphino)ferrocene for the amination of aryl halides and for the first amination of aryl tosylates. The amination of various amines with aryl chlorides, iodides, and tosylates was effected using this novel ligand in combination with a palladium source affording the coupled products in excellent yield. This ligand is exceptionally effective and, though the electron density on the metal helps accelerate the oxidative addition (a necessity for unactivated aryl chlorides), the electron-richness does not negatively impact the reductive elimination since the steric bulk associated with this ligand facilitates this last step of the catalytic cycle (Table 4).1
Table 4.
back to top 
Materials
References
  1. Hamann, B. C.; Hartwig, J. F. J. Am. Chem. Soc. 1998, 120, 7369.