Attention:
Certain features of sigmaaldrich.com will be down for maintenance Saturday afternoon/evening, February 24th starting at 3:00 pm CT until 9:00 pm CT.
Please note that you still have telephone and email access to our local offices. We apologize for any inconvenience.
Fluorous Biphase Catalysis, Kit I: C–C-Coupling
Chemfiles Volume 4 Article 9
The Fluorous Biphase Catalysis Kit for C–C-Couplings contains perfluoro-tagged Pd-catalysts and solvents for up to 10 different catalytic reactions under fluorous biphasic conditions. To get acquainted with this innovative FBC technology our kit provides detailed descriptions of procedures for two C–C-coupling reactions (Suzuki and Stille coupling),[1,2] all substrates and reagents necessary for these two model reactions, and eight additional preparations. Analytical methods and spectra are also given as references. FBC technology allows the catalyst to be easily recovered and to be used for further syntheses.
back to top 
References
  1. Schneider, S.; Bannwarth, W. Helv. Chem. Acta 2001, 84, 1.
  2. Schneider, S.; Bannwarth, W. Angew. Chem., Int. Ed. Engl. 2000, 39, 4142.