Attention:
Certain features of sigmaaldrich.com will be down for maintenance Saturday afternoon/evening, February 24th starting at 3:00 pm CT until 9:00 pm CT.
Please note that you still have telephone and email access to our local offices. We apologize for any inconvenience.
Indanols
Aldrich ChemFiles 2005, 5.4, 7.
Aldrich ChemFiles 2005, 5.4, 7.
1-[(3,5-Di-tert-butyl-2-hydroxybenzylidene)amino]-2-indanol
These chiral imine-ligands have recently been used in the vanadium-catalyzed oxidation of sulfides in conjunction with hydrogen peroxide. Optically active sulfoxides are obtained in high yields and enantioselectivities (Scheme 16).1
Scheme 16
back to top 
Materials
References
  1. Pellotier, B. et al. Synlett 2002, 1055.