Attention:
Certain features of sigmaaldrich.com will be down for maintenance Saturday afternoon/evening, February 24th starting at 3:00 pm CT until 9:00 pm CT.
Please note that you still have telephone and email access to our local offices. We apologize for any inconvenience.
Jørgensen Organocatalyst for Asymmetric Catalysis
Chemfiles Volume 7 Article 5
Recently, the Jørgensen group communicated the use of the prolinol-based silyl ether 1 as an effective organocatalyst for the epoxidation of a, b-unsaturated aldehydes in ethanol and aqueous solutions.1 Hydrogen peroxide was used as the oxidant, and the reactions proceed in moderate to good yields with ee’s up to 96%.
back to top
References
  1. Zhuang, W. et al. Org. Biomol. Chem. 2005, 3, 3883.
back to top
Related Links
Categories