Chemfiles Volume 7 Supplement 1
An exclusive collection of palladium and copper containing perovskite heterogeneous catalysts for use in a range of organic reactions. The LaPCatperovskite catalysts have shown very high catalytic activity in many coupling reactions with a wide variety of substrates.
Cleaner products
typically 10 ppm or less of metal contamination in crude product
Cleaner waste streams
reduced metal losses in LaPCat processes
Faster, efficient processes
equivalent or better than homogeneous catalysts
Improved processes
very low catalyst loadings achieved
Improved cost effectiveness
LaPCat catalysts may be reused in repeat reaction cycles