Metal Scavengers
Chemfiles Volume 7 Supplement 1
QuadraPure and QuadraSil metal scavengers offer a variety of complementary solutions for efficient removal and economic recovery of precious metal contamination from products and process streams (aqueous or organic). The picture shows a typical ‘before-and-after’ scavenger treatment of a metal catalyst solution (initial metal content 1000 ppm) at room temperature.
The QuadraPure™ collection consists of two ranges of functionalized polystyrene beads:
  • Macroporous, low-swell, robust beads for use in manufacturing either in batch or flow processes.
  • Microporous, solvent swellable beads with high capacity intended largely for R&D application.
back to top
The QuadraSil scavengers offer extremely rapid removal of metal catalyst residues. QuadraSil products are based on high-quality spherical silica beads with well defined porosity that can be used in aqueous or organic solution and in batch or flow processes.