Mono-Protected Diaminoalkanes
ChemFiles Volume 3 Article 4
Mono-Boc-protected diaminoalkanes are very useful for the introduction of homobifunctional spacers in many different applications. Additionally, they play an important role as building blocks in the synthesis of polyamines and polyamides.(1-3)
back to top Back to Top
  1. Blunt, J. W. et al. Tetrahedron Lett. 1982, 23, 2793.
  2. Buchardt, O. et al. J. Org. Chem. 1984, 493, 4123.
  3. Kovacs, L.; Hesse, M. Helv. Chim. Acta 1992, 75, 1909.
back to top Back to Top
Related Links