Attention:
Certain features of sigmaaldrich.com will be down for maintenance Saturday afternoon/evening, February 24th starting at 3:00 pm CT until 9:00 pm CT.
Please note that you still have telephone and email access to our local offices. We apologize for any inconvenience.
MRT - Grignard Reaction
By: Matthias Junkers, Aldrich ChemFiles 2009, 9.4, 15.
Aldrich ChemFiles 2009, 9.4, 15.
Benefit from MRT & Flow Chemistry: cost savings combined with accelerated & economic process development for large scale synthesis.
Microreactors also have some simple pragmatic advantages over traditional batch vessels: they take up less space on the factory floor and yet can still be used to make large quantities of product. 2-Benzoyl pyridine is an important building block, with an annual demand of about 15 tons. It is synthesized through a Grignard reaction (Scheme 9), and in a microreactor, this takes less than a minute. The Grignard microreaction is followed by two online quench modules. Precise reaction control leads to a highly pure product making a distillation step unnecessary that is needed for purification of the lower quality batch product.
Scheme 9: Grignard microreaction with two subsequent quench modules
The flow process was first developed with the Sigma-Aldrich Microreactor Explorer Kit (19979) using a 2 mL glass microreactor, which is able to produce up to 1 kg of 2-benzoyl pyridine per day. In a pilot study for the transfer of microreactor technology into large scale production we could show that process economic parameters evaluated with the Sigma-Aldrich Microreactor Explorer Kit (19979) ccould be directly transferred to production in a stainless steel Alfa Laval Art® plate reactor. Thus the rapid flow rates that can be achieved with this reactor allows the continuous production of 200 to 300 of kg 2-benzoyl pyridine per day. All of this from a reactor that requires only 30 x 50 cm of bench space!
back to top Back to Top