Attention:
Certain features of sigmaaldrich.com will be down for maintenance Saturday afternoon/evening, February 24th starting at 3:00 pm CT until 9:00 pm CT.
Please note that you still have telephone and email access to our local offices. We apologize for any inconvenience.
MRT - Ring Expansion
By: Matthias Junkers, Aldrich ChemFiles 2009, 9.4, 13.
Aldrich ChemFiles 2009, 9.4, 13.
Benefit from MRT & Flow Chemistry: safe & scale-independent handling of diazo reagents.
Potentially hazardous reagents like diazo compounds can be handled safely in microreactors because actual reaction volumes are very small. In a procedure developed at Johnson&Johnson ethyl diazoacetate is used in the presence of BF3•Et2O to expand the six-membered ring of N-Boc-4-piperidone.1 Thorough calorimetric investigations showed that this reaction can only be performed safely at small scales (test runs with 70 mg) in batch reactors. Scale-up to kilogram quantities cannot be recommended due to the risk of overheating and excessive nitrogen formation, leading to dangerous over-pressurization of the reaction vessel. The existing small-scale batch protocol was transferred into a flow process without the need of any further process development achieving a throughput of 91 g per hour at 89% yield.
Inspired by the Johnson&Johnson research this procedure was developed further and is used today by Sigma-Aldrich for the catalog production of N-Boc-hexahydro-1H-azepin-4-one (00833). The microreaction is followed by base induced decarboxylation performed in a conventional batch reactor (Scheme 5).
Scheme 5: Michael addition with subsequent Dieckmann condensation and decarboxylation leads to ring expansion (00833)
back to top Back to Top
Materials
References
  1. Zhang, X.; Stefanick, S.; Villani, F.J. Org. Process Res. Dev. 2004, 8, 455.
back to top Back to Top
Related Links