Attention:
Certain features of sigmaaldrich.com will be down for maintenance Saturday afternoon/evening, February 24th starting at 3:00 pm CT until 9:00 pm CT.
Please note that you still have telephone and email access to our local offices. We apologize for any inconvenience.
N-Heterocyclic Carbene-Copper Complexes
By: Josephine Nakhla, Aldrich ChemFiles 2009, 9.2, 16.
N-heterocyclic carbene ligands have proven very popular in the last 20 years. The electronic and steric modularity associated with the resulting complexes have made NHCs obvious candidates when designing new metal complexes for catalysis. Nolan and co-workers are among the pioneers in the use of NHC ligands for catalysis and they have reported the use of Cu-NHCs for a variety of catalytic transformations. Conjugate reduction of a,ß-unsaturated ketones and esters, the hydrosilylation of ketones, the cyclopropanation of terminal alkenes, as well as olefinations, carbene transfer reactions, aziridination of olefins, and methylenation of aldehydes (Scheme 4) are among some examples of the uses of Cu-NHC complexes (specifically (IPr)CuCl) in modern catalysis. Finally, these catalysts are air- and moisture-stable, and they can be used as precursors to synthesize more air-sensitive complexes.1
Scheme 4.
Nolan and co-workers have described the use of the Cu-Imes carbene complex 5 in the olefination of aldehydes (and ketones) in the presence of PPh3, i-PrOH, and TMSCHN2. While similar olefinations have been reported using Wilkinson’s catalyst, the Cu-alternative offers a more economical solution. The functional group compatibility is quite good, allowing for the formation of functionalized aliphatic olefins, dienes, and styrenes containing nitro (notoriously deleterious to these types of reactions), trifluoromethyl, amino, and ester functionalities as well as for heteroaromatic olefins substituted with pyridine, pyrrole, and indole derivatives (Scheme 5). The isopropyl-derived carbene complex 6 was also demonstrated to be quite useful in these types of reactions under slightly modified reaction conditions (Scheme 6).2
Scheme 5.
Scheme 6.
back to top 
Materials
References
  1. (a) Díez-González, S. et al. Organometallics, 2006, 25, 2355. (b) Díez-González, S.; Nolan, S. P. Acc. Chem. Res. 2008, 41, 349. (c) Jurkauskas, V. et al. Org. Lett. 2003, 5, 2417. (d) Lebel, H. et al. J. Org. Chem. 2007, 72, 144. (e) Trost, B. M.; Dong, G. J. Am. Chem. Soc. 2006, 128, 6054. (f) Fructos, M. R. et al. J. Am. Chem. Soc. 2004, 126, 10846. (g) Kaur H. et al. Organometallics 2004, 23, 1157. (h) Díez-González, S.; Nolan, S. P. Aldrichimica Acta 2008, 41, 43.
  2. Lebel, H. et al. J. Org. Chem. 2007, 72, 144.