Attention:
Certain features of sigmaaldrich.com will be down for maintenance Saturday afternoon/evening, February 24th starting at 3:00 pm CT until 9:00 pm CT.
Please note that you still have telephone and email access to our local offices. We apologize for any inconvenience.
Pd(II) EnCat™
Chemfiles Volume 7 Supplement 1
Pd(II) EnCatcovers a versatile range of immobilized palladium acetate catalysts with or without activating ligands contained in a robust, porous polyurea matrix which allow easier, faster and cleaner C–C coupling processes to be developed.
Cleaner products
typically less than 100 ppm Os in crude product
Cleaner waste streams
reduced metal losses in Pd(II) EnCat processes
Faster, efficient processes
EnCat beads filter easily
No plant contamination
metal & ligands are trapped within the beads
Improved processes
high activity in many types of coupling reactions
Process intensification
can be used in batch & flow processes