Attention:
Certain features of sigmaaldrich.com will be down for maintenance Saturday afternoon/evening, February 24th starting at 3:00 pm CT until 9:00 pm CT.
Please note that you still have telephone and email access to our local offices. We apologize for any inconvenience.
Polymer-Bound Lewis Acids
Chemfiles Volume 5 Article 11
Lewis acids are important catalysts for many types of transformations in organic chemistry. Lanthanide salts serve as mild alternatives to traditional hard Lewis acids. Reactions catalyzed by the polymerbound lanthanide Lewis acids include acetalization, aldol reaction and allylation of aldehydes, nucleophilic addition to imines, aza- Diels–Alder reactions, and ring opening of epoxides.1 Polymer-bound reagents react the same as the solution-phase equivalent and have the added advantage of ease of handling, simplified reaction workup and, in some cases, recyclability.
back to top
Ytterbium(III) Polystyrenesulfonate
Typical Experimental Procedure for Ring Opening of Epoxides
Styrene oxide (0.12 mL, 1 mmol) and ytterbium(III) polystyrenesulfonate polymer-bound (100 mg, 0.056 mmol) were mixed in methanol (1 mL), and stirred at room temperature overnight (16 h). TLC showed that the reaction went to completion. The reaction was filtered and the resin washed with methanol (2 × 1 mL). The combined solution phases were evaporated to give 2-methoxy-1- phenylethanol a colorless oil (151 mg, 100%).
back to top
Scandium(III) Bis(trifluoromethanesulfonate), Polymer-Bound
One of the important transition metal Lewis acid catalysts is scandium(III) bis(trifluoromethanesulfonate), which is widely used in carbon–carbon bond and carbon–heteroatom bond formation.2
Typical Experimental Procedure for Addition of Allyl Group to Esters
Ethyl benzoylformate (178 mg, 1 mmol), tetraallyltin (282 mg, 1 mmol) and scandium bis(trifluoromethanesulfonate), polymerbound(10 mg, 0.0068 mmol) were mixed in water (6 mL). The mixture very quickly became cloudy and was stirred at room temperature overnight. Then ether (20 mL) was added and the mixture was passed through a silica gel pad. The organic layer was separated, dried, and evaporated to afford a white solid (205 mg, 95% yield). The structure was confirmed by 1H NMR.
back to top
Materials
References
  1. Dondoni, A.; Massi, A. Tetrahedron Lett. 2001, 42, 7975. Yu, L.; Chen, D.; Li, J.; Wang, P. G. J. Org. Chem. 1997, 62, 3575.
  2. Kobayashi, S.; Sugiura, M.; Kitagawa, H.; Lam, W. W. L. Chem. Rev. 2002, 102, 2227. Kobayashi, S.; Kitagawa, H.; Matsubara, R. J. Comb. Chem. 2001, 3, 401.