Potassium 2-isocyanoacetate: Quenching Reagent for Metathesis
By: Dr. Mark Redlich, Chemfiles Volume 10 Article 1
Diver and co-workers recently described an efficient metathesis clean-up procedure using potassium 2-isocyanoacetate,1 which addresses the challenges associated with removal of the ruthenium at the end of a metathesis reaction. Upon quenching with the polar isocyanide, a benzylidene insertion of the mesityl group occurs, thus destroying the catalyst and imparting increased polarity for easy removal (Scheme 7). Potassium 2-isocyanoacetate was successfully used to quench a range of metathesis reactions and the authors demonstrated it can be employed with various highly utilized Grubbs' complexes.
Scheme 7
Scheme 7: (705438)
back to top
  1. Galan, B. R. et al. Org. Lett. 2007, 9, 1203.