Potassium Vinyltrifluoroborate as a Versatile Dianion Precursor
By: Aaron Thornton, Chemfiles Volume 11 Article 1
Molander and co-workers have developed a powerful strategy for the production of a unique 1,2-dianion equivalent using potassium vinyltrifluoroborate.1 This useful organotrifluoroborate undergoes selective hydroboration with 9-BBN to generate (3), a 1,2-dianion equivalent that can then undergo a variety of selective transformations (Scheme 1).
Scheme 1: Hydroborated intermediate (3) as a 1,2-dianion equivalent.(655228)
This versatile 1,2-dianion equivalent undergoes sequential Suzuki-Miyaura cross-coupling with a range of organic electrophiles, including aryl-, heteroaryl-, and alkenyl halides (Table 1).
Table 1: Sequential Suzuki-Miyaura cross-couplings to build molecular complexity from potassium vinyltrifluoroborate.
back to top 
  1. Molander, G. A.; Ellis, N. Acc. Chem. Res. 2007, 40, 275.
  2. Molander. G. A.; Sandrock, D. L. Org. Lett. 2009, 11, 2369.