Attention:
Certain features of sigmaaldrich.com will be down for maintenance Saturday afternoon/evening, February 24th starting at 3:00 pm CT until 9:00 pm CT.
Please note that you still have telephone and email access to our local offices. We apologize for any inconvenience.
Proline Derivatives
By: Matthias Junkers, Aldrich ChemFiles 2008, 8.7, 8.
Aldrich ChemFiles 2008, 8.7, 8.
Proline is a non-polar, natural amino acid that forms a tertiary amide when incorporated into peptides. Thus, it is the only proteinogenic amino acid that does not act as a hydrogen bond donor in a peptide chain. Proline is known as a classical breaker of both the α-helical and β-sheet secondary structures in proteins and peptides, and it plays a crucial role in protein folding. Synthetic proline derivatives, mimetics and analogs offer further options to tune the biological, pharmaceutical, or physicochemical properties of peptides and proteins. In recent years proline derivatives and analogs have also found increasing popularity as organocatalysts in asymmetric synthesis.1
back to top Back to Top
Materials
References
  1. (a) Vignola, N.; List, B. J. Am. Chem. Soc. 2003, 125, 450. (b) Dalko, P.I.; Moisan, L.; Angew. Chem. Int. Ed. 2004, 43, 5138.
back to top Back to Top
Related Links