Attention:
Certain features of sigmaaldrich.com will be down for maintenance Saturday afternoon/evening, February 24th starting at 3:00 pm CT until 9:00 pm CT.
Please note that you still have telephone and email access to our local offices. We apologize for any inconvenience.
Propargyl Claisen Rearrangement
By: Josephine Nakhla, Chemfiles Volume 10 Article 4
Toste and coworkers reported the use of the gold catalyst [(Ph3PAu)3O]BF4 for the rapid two-step, one-pot sequence of a Claisen rearrangement of a propargyl vinyl ether followed by a reduction of the resulting aldehyde functionality to provide a variety of homoallenic alcohols. The reactions are generally high yielding and the robust catalyst system is able to induce almost complete chirality transfer (in most cases, ee’s were >90%). Low catalyst loadings (1 mol %) and substitution at the alkyne is well tolerated generating the desired allenes in high yields (Scheme 3).
Scheme 3: Preparation of homoallenic alcohols via Claisen rearrangement/ reduction sequence
back to top
References
  1. Sherry, B. S.; Toste, F. D. J. Am. Chem. Soc. 2004, 126, 15978.