Rink Acid Resins
ChemFiles Volume 3 Article 4
Rink acid resins are extremely acid-sensitive supports. Protected peptide acid fragments can be obtained without any loss of sidechain protection functionality by applying 10% acetic acid in dichloromethane. Consecutive coupling steps during chain elongation should be carried out under basic conditions to prevent any premature cleavage of peptide chains.
back to top 
  1. Rink, H., Tetrahedron Lett. 1987, 28, 3787.