Rovis Triazolium Catalyst
ChemFiles 2006, 6.4, 14.
Rovis has demonstrated that triazolium salt (667080) in the presence of a base can act as an N-heterocyclic carbene organocatalyst in highly enantioselective intramolecular Stetter reactions. The Stetter reaction (a conjugate addition of an aldehyde to an α,β-unsaturated compound) is a superb method for construction of 1,4-dicarbonyl compounds bearing quaternary stereocenters (Scheme 16).1
Scheme 16.
back to top
Materials
References
  1. Kerr, M. S.; Rovis, T. J. Am. Chem. Soc. 2004, 126, 8876.