Attention:
Certain features of sigmaaldrich.com will be down for maintenance Saturday afternoon/evening, February 24th starting at 3:00 pm CT until 9:00 pm CT.
Please note that you still have telephone and email access to our local offices. We apologize for any inconvenience.
Suzuki Coupling of Alkyl Chlorides
By: Josephine Nakhla, Aldrich ChemFiles 2009, 9.2, 12.
Aldrich ChemFiles 2009, 9.2, 12.
Gonzalez-Bobes and Fu reported the use of NiCl2•glyme in the presence of prolinol in the previously unprecedented Suzuki reactions of unactivated secondary alkyl chlorides. Both primary, secondary, cyclic and acyclic alkyl chlorides can be utilized in this transformation, as well as electron-rich and electron-poor arylboronic acids (Table 5).1
Table 5.
back to top 
Materials
References
  1. Gonzalez-Bobes, F.; Fu, G. C. J. Am. Chem. Soc. 2006, 128, 5360.