Symmetrical Rf-tagged 1,3-Diketone Ligand
Chemfiles Volume 4 Article 9
Triacontafluoro-8,10-heptadecanedione has been described as an symmetrical perfluorinated ligand for transition metal catalyzed oxidations in perfluorinated solvents.[1]
back to top 
  1. Klement, I.; Lütjens, H.; Knochel, P. Angew. Chemie, Int. Ed. Engl. 1997, 26, 1454