Attention:
Certain features of sigmaaldrich.com will be down for maintenance Saturday afternoon/evening, February 24th starting at 3:00 pm CT until 9:00 pm CT.
Please note that you still have telephone and email access to our local offices. We apologize for any inconvenience.
Symphos
By: William Sommer, Aldrich ChemFiles 2007, 7.10, 11.
Aldrich ChemFiles 2007, 7.10, 11.
A new class of phenyl-backbone derived P,O-ligands (Symphos) were developed by Symyx Technologies in collaboration with professor Fu and coworkers.1 These new ligands proved to be suitable for general palladium/ligand catalyzed Suzuki transformations of aryl halides. This new system demonstrated its efficiency by catalyzing the cross-coupling of aryl boronic acids and aryl chlorides containing electron-poor and electron-rich substituents with good overall yields (Scheme 1). The catalysis was carried out with Pd(dba)2 and Symphos with CsF in toluene at 105 °C with yields of up to 96% (Table 1).
Scheme 1
Table 1
back to top Back to Top
Materials
References
  1. Bei, X et al. J. Org. Chem. 1999, 64, 6797.
back to top Back to Top
Related Links