Attention:
Certain features of sigmaaldrich.com will be down for maintenance Saturday afternoon/evening, February 24th starting at 3:00 pm CT until 9:00 pm CT.
Please note that you still have telephone and email access to our local offices. We apologize for any inconvenience.
Unnatural Amino Acids
By: Matthias Junkers, Aldrich ChemFiles 2007, 7.2, 12.
Aldrich ChemFiles 2007, 7.2, 12.
Unnatural amino acids are utilized as chiral building blocks, conformational constraints, molecular scaffolds, or pharmacologically active compounds. They represent a nearly infinite array of diverse structural elements for the development of new therapeutic drugs. Small-molecule combinatorial libraries containing unnatural amino acid residues already show remarkable impact on drug discovery processes. Novel short-chain peptide ligand mimetics with both enhanced biological activity and improved proteolytic resistance are drug candidates in today’s R&D pipelines of pharmaceutical companies. Sigma-Aldrich is pleased to introduce 40 new additions to its unique and broad portfolio of more than 700 unnatural amino acids.
back to top Back to Top