Attention:
Certain features of sigmaaldrich.com will be down for maintenance Saturday afternoon/evening, February 24th starting at 3:00 pm CT until 9:00 pm CT.
Please note that you still have telephone and email access to our local offices. We apologize for any inconvenience.
Unsaturated Amino Acids
By: Matthias Junkers, Chemfiles Volume 10 Article 4
Recent developments in Chemical Biology research have increased the demand of amino acid building blocks with unsaturated side chains. Alkyne moieties can be used in bioorthogonal synthesis strategies to form hybrid structures, introduce chemical probes into biomolecules, or link large fragments with each other. The most prominent technique relies on the Huisgen dipolar cycloaddition reaction between an azide and an alkyne.
Olefi n moieties open amino acids and peptides to metathesis reactions and a full range of other bioorthogonal synthesis routes. Olefi n metathesis is a key to the production of hydrocarbon stapled peptides.1 Stapled peptides are currently in discussion as a new class of superpotent drugs or magical bullets promising to make peptide α-helices more potent and cell permeable by locking them in the most active conformation.2
back to top
References
  1. Walensky, L.D. et al. Science 2004, 305, 1466.
  2. Kritzer, A.K. Nature Chemical Biology 2010, 6, 566.