Potential Application for 3-Methyl-1,2-Cyclohexanedione, FG
Available in Halal
This product’s sweet roasted coffee bean notes give it definite application in coffee flavors. It also would be very useful in a range of toasted and roasted flavors such as burnt sugar, caramel, butterscotch, toffee, molasses and toasted bread flavors. It would also add interesting character to roasted sesame and maple flavors.
Key Organoleptic Characteristics   Roasted coffee, caramellic, maple and toasted notes
Potential Applications Toasted or roasted flavors
Natural Occurrence Coffee
Aroma Characteristics @1.0% Ethanol. Sweet burnt caramellic furanone like with coffee, maple and toasted bready notes with nuances of brown sugar, molasses and cotton candy.
Taste Characteristics @<10ppm. Sweet burnt caramellic, roasted coffee depth notes with maple and furaneol-like brown sugar-like notes.
Product Number W330507