Potential Application for Butyl 2-Methylbutyrate
Available in Natural (US) & Kosher
This product finds application in both flavor and fragrances. It can be utilized as a masking agent for sulfurous odors or other volatile malodors. It is more commonly found in flavor formulations for apple, banana and other various fruit complexes.
Key Organoleptic Characteristics   Sweet fruity, juicy tropical, ripe tutti-frutti
Potential Applications Apple, strawberry, white grape, mango, guava, papaya, pineapple, cherry, honey, apricot and peach
Natural Occurrence Apple, apricot, banana, sweet cherry, melon, mountain papaya, plum, starfruit, strawberry
Aroma Characteristics @1.0% in ethanol. Sharp green, fresh fruity with tropical notes of mango, peach, honey and cherry
Taste Characteristics @5-10 ppm. Fruity apple, white grape, plum, strawberry with tropical nuances of mango, guava and papaya
Product Number W339315