Potential Application for D-Dihydrocarvone
Available in Mixture of Isomers, Natural & Kosher
D-Dihydrocarvone finds good use as a spearmint/mint flavor enhancer. It would find application in chewing gum, oral care and analgesic cough syrup formulations. It is usually employed in combination with other mint and herbal oils and also works well with high potency coolants.
Key Organoleptic Characteristics   Ethereal, weedy, spicy, caraway with minty and camphoreous nuances
Potential Applications Oral care products; confections; spice blends and flavored tea
Natural Occurrence Caraway seed, celery seed, dill, patchouli oil china, spearmint
Aroma Characteristics @1.0% in ethanol. Ethereal, herbal spicy, woody, caraway with minty spearmint and weedy nuances
Taste Characteristics @5-10 ppm. Sweet creamy, candy-like with good mouth feel
Product Number W356511