Potential Application for Delta Undecalactone Mixed Isomers, FG
Available in Natural & Kosher
This ingredient is versatile profile offers creamy apricot and peach notes. The creamy effect of this product finds use in ice creams and beverages. This product has also been applied in floral applications most notably magnolia or gardenia.
Key Organoleptic Characteristics   Sweet, lactonic, fruity apricot and peach with rich, waxy, creamy dairy notes
Potential Applications Fruit flavors such as apricot, peach, tropical fruits such as mango and papaya, dairy flavors such as fresh and cooked butter, ghee, buttermilk, ice cream, fluid milk, condensed milk, coffee creamers, nut flavors such as macadamia and pecan
Natural Occurrence Blackberry, coconut, cream, milk, heated butter
Aroma Characteristics @1.0% in ethanol. Sweet lactonic, fruity peach and apricot with waxy creamy dairy depth notes
Taste Characteristics @1-5 ppm. Creamy dairy, waxy lactonic, creamy with fruity peach, apricot and tropical richness with buttery nutty nuances reminiscent of pecan and macadamia
Product Number W329415