Potential Application for Ethyl Phenylacetate, FG
Available in Natural & Kosher
Ethyl phenylacetate’s impactful, honey-like aromas are useful in fruity applications such as: cherry, apricot and peach; along with its more traditional usage in honey flavors. It also works well in perfume formulations, where it offers subtle nuances appropriate for the development of an oriental note.
Key Organoleptic Characteristics   Honey, sweet, anise, apple, apricot, cherry, chocolate, tropical, melon, raspberry, rose, wine-like, balsamic cocoa-like with molasses and yeasty nuances.
Potential Applications Cherry, apricot and peach, along with its more traditional usage in honey flavors
Natural Occurrence Apple, beer, brandy, bread, cider, grapefruit juice, guava fruit
Aroma Characteristics @1.0%. Sweet honey, floral rose, slightly fruity with black licorice, tobacco and dried fruit nuances of fig, date and raisin
Taste Characteristics @10 ppm. Sweet honey, balsamic floral with cocoa and dried fruit nuances of date, fig and raisin
Product Number W245230