Potential Application for Methional
Available in Natural & Kosher
Methional supplies the necessary earthy mustiness found in cheddar cheese and various other mold ripened cheeses. It carries the typical array of tomato and potato flavor characteristics. It provides depth to chicken bouillon flavors.
Key Organoleptic Characteristics   Earthy, vegetative potato and tomato
Potential Applications Fresh tomato; baked, mashed and fried potato; cheddar cheese and various nut and malted milk applications
Natural Occurrence Beans, beer, bread, cheddar cheese, gorgonzola cheese, cooked chicken, ham, buckwheat honey, macadamia nut, mushroom, peanut, baked potato, potato chip, pumpkin, tomato
Aroma Characteristics @1.0% in ethanol. Vegetative potato, tomato, mushroom, metallic, with nuances of cheddar cheese and nut
Taste Characteristics @0.5-1.0 ppm. Vegetative tomato, mushroom, earthy, mashed, baked and fried potato-like with savory meaty, cheese and fruity nuances
Product Number W274711