KitAlysis™ C-N (Buchwald-Hartwig) High-Throughput Screening Kit
KitAlysis header
Materials Included in your KITALYSIS-CN-1KT High-Throughput Screening Kit
Contents in each of the 4 individually sealed Mylar (foil bags):
  • 24 (12 x2) pre-weighed catalysts in glass vials loaded with stir bars, topped with cap mat.
The screening sets come pre-loaded with 1 µmol of catalyst in each vial according to the following design.
Mouse over image to select.
Description Product No. Vial
BrettPhos Pd G3 761605 A1, C1
CPhos Pd G3 763004 A2, C2
DavePhos Pd G3 804959 A3, C3
P(tBu3) Pd G2 756482 A4, C4
J009 Pd G3 747130 A5, C5
MorDalPhos Pd G3 792357 A6, C6
BINAP Pd G3 804967 B1, D1
RuPhos Pd G2 753246 B2, D2
SPhos Pd G2 753009 B3, D3
tBuXPhos Pd G3 762229 B4, D4
XantPhos Pd G3 763039 B5, D5
XPhos Pd G3 763381 B6, D6
  • Cesium Carbonate: ground and sieved in 2 pre-weighed vials with product labels and lids
  • 2 empty reaction vials with lids
All contents in the foil bag are weighed, plated, packed, and sealed in a glove box under nitrogen.
Ampule Boxes:
  • Dioxane: 10 x 2 mL of degassed, anhydrous Aldrich® Sure/Seal™
  • DMAc: 10 x 2 mL of degassed, anhydrous Aldrich® Sure/Seal™
  • NaOt-Bu Solution: 4 x 0.5 mL degassed anhydrous Sure/Seal™ 2 M THF solution
Internal Standard:
  • Biphenyl (30 mg). To be added to the reaction during the work-up. Recipe for the work-up with the internal standard can be found within the hyperlinked screen types in the "Kit Design" section below.
KitAlysis™ 24-well Reaction Block Replacement Films-4EA
  • Pack of 4 enables a new film to be used with each kit ensuring a tight, cross-contaminate-free seal every time.
Stir Bars-8 individually packed
  • Fit perfectly into supplied vials to ensure proper stirring of substrate mixtures.
Additional Recommended Materials (Sold Separately)
  • Z688711: 96-well plate for automated HPLC analysis
  • C1111: 96-well plate cap mat for automated HPLC analysis
  • Z683809: 10-100 uL pipette
  • Z640220: 2-200 uL pipette tip refill
  • Z683825: 100-1000 uL pipette
  • Z640247: 50-1000 uL pipette tip refill
  • Z192570 ALDRICH: 2 inch needles for ease in ampule solvent extraction
  • Z230723 ALDRICH: 1 mL needle for accurate solvent volume extraction from ampule
Kit Design
C-N (Buchwald-Hartwig) cross-coupling kit was designed to provide the best possible chance of success and is run with:
  • 10 umol aryl halide
  • 15-20 umol amine (primary, secondary), use 20 umol if the material is not too precious
  • 1 umol catalyst per vial (1:1 Pd:Ligand); 12 in this system
  • 30 umol base (NaOt-Bu or Cs2Co3)
  • 0.1 M solvent concentration (Dioxane or DMAc)
The C-N (Buchwald-Hartwig) cross-coupling kit allows the end user ultimate control and flexibility based upon their specific chemical system. For each kit, there are four possible screens that can be run based upon the substrates you provide:
The following guidelines can be used to determine your starting point:
Substrate characteristics: Screen Type: Attempt:
base sensitive C-N: Two Solvents, Weak Base first
base tolerant C-N: Two Solvents, Strong Base first
base tolerant C-N: Dioxane, Two Bases second: based upon substrate solubility
base tolerant C-N: DMAc, Two Bases second: based upon substrate solubility
Not Sure C-N: Two Solvents, Strong Base first
Not Sure C-N: Two Solvents, Weak Base second
The kit was purposefully designed to be modular, meaning that you can run 4 different kits, 4 identical kits, one of each, or any combination. All of the supplies can be mixed and matched to meet your needs.
Step-by-step user guide and an excel sheet for stock solution recipes that can be downloaded within each step-by-step user guide hyperlinked above.
Use of the Provided Tools
Multiple tools have been created to ensure your success with kit set up. Start with the more detailed guide to ensure you are comfortable with all of the steps before using the quick guides on the excel worksheet. Remember that while the technique is new, it is still organic chemistry and so the steps will seem easy once you try just one kit. It is just a new way of approaching something you are already very good at.
Detailed Set-Up User Guide:
Designed for the first time user and should be read completely before getting started. Best if used in conjunction with the video as not all steps are outlined in the video in great detail. This guide includes trouble-shooting tips, how-to’s for the Labware, and work-up recipes with procedures. Everything you need to set up a kit with confidence every time.
Excel Sheet:
Each sheet is designed to be used with the specific experimental design chosen by you depending upon the attributes of your substrates. The downloadable excel files are specific to the kit being run and can be found within each Step-by-Step User Guide. They have the following features:
  • Calculations for substrate recipies depending upon the molecular weight of your substrates
  • Quick directions for the more avid user
  • Print button to allow you to take recipe to the lab
  • Contain all info such that they can be can be saved as a pdf and appended to ELN for experimental information