KitAlysis™ High-Throughput Metal Scavenger Reaction Screening Kit
KitAlysis header
Materials Included in your KITALYSIS-SCAV-2PK High-Throughput Screening Kit
Contents in each of the 2 individually sealed Mylar (foil bags):
  • 24 (12 x 2) pre-weighed scavangers / resins in glass vials loaded with stir bars, topped with cap mat.
The screening sets come pre-loaded with 12 scavengers / resins (10mg or 50mg) in each vial according to the following design.
Mouse over image to select.
KitAlysis
Name Product Number Vials Loading
Cysteine functionalised Silica Gel 900677 A1
C1
10mg
50mg
N-Propyldiethanolamine-functionalized silica gel 900676 A2
C2
10mg
50mg
triaminetetraacetate Na salt silica gel 900674 A3
C3
10mg
50mg
triaminetetraacetic acid-functionalised silica gel 900675 A4
C4
10mg
50mg
DMT-functionalized silica gel 900680 A5
C5
10mg
50mg
3-(diethylenetriamino)propyl-functionalized silica gel 537926 A6
C6
10mg
50mg
Mixture of metal scavengers on silica gel 900673 B1
D1
10mg
50mg
DOTA-functionalized silica gel 900679 B2
D2
10mg
50mg
AMPA-functionalized silica gel 900678 B3
D3
10mg
50mg
3-(Ethylenediamino)propyl functionalized silica gel 537918 B4
D4
10mg
50mg
3-(1-thioureido)propyl functionalized silica gel 569879 B5
D5
10mg
50mg
3-Mercaptopropyl-functionalized silica gel 538086 B6
C6
10mg
50mg
All contents in the foil bag are weighed, plated, packed, and sealed in a glove box under nitrogen.
Additional Recommended Materials (Sold Separately)
  • Z683809: 10-100 uL pipette
  • Z640220: 2-200 uL pipette tip refill
  • Z683825: 100-1000 uL pipette
  • Z640247: 50-1000 uL pipette tip refill
  • Z192570 ALDRICH: 2 inch needles for ease in ampule solvent extraction
  • Z230723 ALDRICH: 1 mL needle for accurate solvent volume extraction from ampule
  • 270997 : Dichloromethane (anhydrous)
  • 270989 : Ethyl acetate (anhydrous)
  • 186562 : Tetrahydrofuran (anhydrous)
  • 227056 : N,N-Dimethylformamide (anhydrous)
  • 244511 : Toluene (anhydrous)
Kit Design
The KitAlysis™ Metal Scavenger kit was designed to quickly allow you to optimize scavenger choice in removing excess metals from reactions or isolated product. Specifically chosen by chemistry experts, each of the twelve scavengers is provided in 10 mg and 50 mg amounts to quickly assess not only the optimal scavenger, but the optimal w/w% of scavenger to substrate as well.
Use of the Provided Tools
Multiple tools have been created to ensure your success with kit set up. Remember that while the technique is new, it is still organic chemistry and so the steps will seem easy once you try just one kit. It is just a new way of approaching something you are already very good at.
Detailed Set-Up User Guide:
Step-by-Step Guide for KitAlysis™ High-Throughput Metal Scavanger Reaction Screening Kit