Pressure-Temperature Nomograph
Slider A is currently locked as the pivot. Slider B is currently locked as the pivot. Slider C is currently locked as the pivot.
A
Observed Boiling Point at Pressure "P" (mmHg)
B
Boiling Point Corrected
at 1 atm (760 mmHg)
C
Pressure "P" (mmHg)
1 atm = 760 mmHg
To find a boiling point under a set vacuum
Chart B - Enter the known boiling point (or slide the bar) and click the lock.
Chart C - Select the pressure you are working at (Torr or mmHg) and click the lock.
To find a corrected boiling point @760 mm
Chart A - Enter the observed boiling point (or slide the bar) and click the lock.
Chart C - Select the pressure you are working at (Torr or mmHg) and click the lock.