Colby Group - Professor Product Portal
Professor Product Portal Index
Professor David Colby
Prof. David Colby's laboratory is developing synthetic methods to enable the efficient modification of complex molecules, such as natural products, as well as the production of fluorinated compounds. They have focused on understanding how to cleave bonds in order to remodel structures, and their first major discovery is that the release of trifluoroacetate is a powerful approach to break carbon–carbon bonds. This strategy has enabled the invention of new reagents and methods for the synthesis of fluorinated products for drug discovery and for exploring medicinal chemistry.
Colby Group website
For applicable product information, please click below:
For recent articles from the Colby Laboratory
Products available at Aldrich from the Colby Laboratory
Mark V Riofski, Allison D Hart, David A Colby
Organic Letters 2013-01-04
A powerful, new reagent, an amidinate salt of hexafluoroacetone hydrate, is an air-stable salt that can be used for the preparation of fluorinated organic molecules. Nucleophilic trifluoromethylation reactions are demonstrated following the base-promoted release of trifluoroacetate. This reagent is soluble in many polar organic ...Read More