Attention:
Certain features of sigmaaldrich.com will be down for maintenance Saturday afternoon/evening, February 24th starting at 3:00 pm CT until 9:00 pm CT.
Please note that you still have telephone and email access to our local offices. We apologize for any inconvenience.
Ellman Group – Professor Product Portal
Professor Product Portal Index
Professor Jonathan A. Ellman
The Ellman group has participated in the development of a variety of C-H functionalization methods.  An electron rich phosphine ligand has proven to be very useful for a variety of Rh(I)-catalyzed C-C bond forming reactions applicable to heterocycle synthesis as exemplified in the recent Science paper “Proton Donor Acidity Controls Selectivity in Nonaromatic Nitrogen Heterocycle Synthesis.” Another useful ligand developed for the highly functional group compatible direct arylation of nitrogen heterocycles is described in a 2008 J. Am. Chem. Soc.  paper “Rh(I)-Catalyzed Arylation of Heterocycles via C-H Bond Activation: Expanded Scope through Mechanistic Insight.”
The Ellman group also developed the chiral amine reagent tert-Butanesulfinamide, which is extensively used in academics and industry for the asymmetric synthesis of amines. A comprehensive survey of tert-Butanesulfinamide methods and applications up through 2009 is provided in the 2010 Chemical Reviews article, “Synthesis and Applications of tert-Butanesulfinamide.”
Ellman Group website
For applicable product information, please click below:
For recent articles from the Ellman Laboratory
Products available at Aldrich from the Ellman Laboratory
Simon Duttwyler, Shuming Chen, Michael K Takase, Kenneth B Wiberg, Robert G Bergman, Jonathan A Ellman
Science 2013-02-08
Piperidines are prevalent in natural products and pharmaceutical agents and are important synthetic targets for drug discovery and development. We report on a methodology that provides highly substituted piperidine derivatives with regiochemistry selectively tunable by varying the strength of acid used in the reaction. Readily a...Read More
MaryAnn T Robak, Melissa A Herbage, Jonathan A Ellman
Chemical Reviews 2010-06-09
Jared C Lewis, Ashley M Berman, Robert G Bergman, Jonathan A Ellman
Journal of the American Chemical Society 2008-02-27
A practical, functional group tolerant method for the Rh-catalyzed direct arylation of a variety of pharmaceutically important azoles with aryl bromides is described. Many of the successful azole and aryl bromide coupling partners are not compatible with methods for the direct arylation of heterocycles using Pd(0) or Cu(I) catal...Read More