Laszlo Kurti Group - Professor Product Portal
Professor Product Portal Index
Professor Andrei Yudin
Photo Credit:
Jeff Fitlow, employed by Rice University
Professor Laszlo Kurti
Amines and their derivatives are ubiquitous substances since they make up the overwhelming majority of drug molecules, agrochemicals as well as many compounds that are produced by plants and living organisms (i.e., natural products). Aromatic amines appear as substructures in more than one third of drug candidates while aziridines, in which the nitrogen atom is bridged between two carbon atoms, are high-reactive and versatile building blocks for a large variety of functionalized amines. Not surprisingly, organic chemists spend a considerable amount of their time with the synthesis and late-stage functionalization of amines that serve as key chemical building blocks for the preparation of biologically active compounds, especially in medicinal chemistry. There is an urgent need for the development of novel carbon-nitrogen bond-forming methods and reagents that expand the toolbox of synthetic organic chemists and enable the environmentally friendly construction of complex molecular structures using the fewest number of chemical steps and generating the least amount waste. A highly attractive, but currently underdeveloped, approach is the utilization of weak bonds as a driving force to achieve the rapid formation of much stronger bonds under mild conditions.
The Kürti lab has been exploring several fundamentally new strategies for the transition-metal-free direct: (i) primary amination of arylmetals such as aryl Grignard reagents and arylboronic acids; (ii) intramolecular C(sp2)-H amination of arenes; (iii) double arylation of a suitable nitrogen linchpin reagents to afford N,N-diarylamines. We have also discovered, in collaboration with the Falck (UTSW) and Ess labs (BYU), the Rh-catalyzed direct N-H/N-alkyl aziridination of olefins as well as the primary (-NH2) and NH-alkyl amination of arenes, transformations that eluded synthetic chemists for decades. In-depth experimental and computational studies have already identified the critical factors required for efficient olefin NH- and N-alkyl aziridination as well as direct arene primary amination and led to the development of practical and chemoselective aminating agents.
Kürti Group Website
Direct and Stereospecific Synthesis of Unprotected N-H and N-Me Aziridines from Olefins
Rapid Heteroatom-Transfer to Arylmetals Utilizing Multifunctional Reagents Scaffolds
Practical Singly and Doubly Electrophilic Aminating Agents: A New, More Sustainable Platform for Carbon-Nitrogen Bond-Formation
For applicable product information, please click below:
For recent articles from the Kurti Laboratory:
Products available at Sigma-Aldrich in collaboration with the Kurti Laboratory
JL Jat,MP Paudyal,H Gao,QL Xu,M Yousufuddin,D Devarajan,DH Ess,L Kürti,JR Falck
Science (New York, N.Y.) 2014-01-03
Despite the prevalence of the N-H aziridine motif in bioactive natural products and the clear advantages of this unprotected parent structure over N-protected derivatives as a synthetic building block, no practical methods have emerged for direct synthesis of this compound class from unfunctionalized olefins. Here, we present a Read More
H Gao,Z Zhou,DH Kwon,J Coombs,S Jones,NE Behnke,DH Ess,L Kürti
Nature chemistry 2017-06-23
Arylmetals are highly valuable carbon nucleophiles that are readily and inexpensively prepared from aryl halides or arenes and widely used on both laboratory and industrial scales to react directly with a wide range of electrophiles. Although C-C bond formation has been a staple of organic synthesis, the direct transfer of primaRead More
PV Kattamuri,J Yin,S Siriwongsup,DH Kwon,DH Ess,Q Li,G Li,M Yousufuddin,PF Richardson,SC Sutton,L Kürti
Journal of the American Chemical Society 2017-06-26
Given the importance of amines in a large number of biologically active natural products, active pharmaceutical ingredients, agrochemicals, and functional materials, the development of efficient C-N bond-forming methods with wide substrate scope continues to be at the frontier of research in synthetic organic chemistry. Here, weRead More
Related Links