Shibasaki – Professor Product Portal
Professor Product Portal Index
Professor Abigail Doyle Professor Masakatsu Shibasaki
Professor Shibasaki's research focuses on the development of novel cooperative asymmetric catalytic systems that allowed for streamlined synthesis of enantioenriched high-value chiral building blocks. The power of cooperativity, i.e. simultaneous activation of nucleophiles and electrophiles by designed multiple catalytic functions, engages low-reactive substrates in the reaction of interest without the aid of stoichiometric reagents. Rare earth metal catalysis developed in our research group is robust and powerful to operate routine asymmetric syntnthesis with a wide range of reaction portfolio, thanks to these metals' ligand-tuned Lewis acidity, high coordination number, variable coordination mode. One note: the word 'rare' in rare earth metals is reminiscent that they are rare elements. But, most of them are abundant in Earth's crust and waiting for being used by scientists to contribute to our society.
Shibasaki Group Website
For applicable additional information, please see below:
For recent articles from the Shibasaki Laboratory:
Product available from the Shibasaki Laboratory:
Opie CR, Noda H, Shibasaki M, Kumagai N.
Chemistry. 2019 Mar 27;25(18):4648-4653. doi: 10.1002/chem.201900715. Epub 2019 Mar 8.
The B3 NO2 six-membered heterocycle (1,3-dioxa-5-aza-2,4,6-triborinane=DATB), comprising three different non-carbon period 2 elements, has been recently demonstrated to be a powerful catalyst for dehydrative condensation of carboxylic acids and amines...Read More
Shuji Masumoto, Hiroyuki Usuda, Masato Suzuki, Motomu Kanai, and Masakatsu Shibasaki*
J. Am. Chem. Soc., 2003, 125 (19), pp 5634–5635
A new method for the catalytic enantioselective Strecker reaction (cyanation) of N-diphenylphosphinoyl ketoimines is described. The asymmetric catalyst is a chiral gadolinium complex prepared from Gd(OiPr)3 and the d-glucose-derived ligand 3 in a 1:2 ratio...Read More