Attention:
We are currently upgrading the certain systems that support this website.  You may experience some functionality loss, though we expect the site to be fully functional by 9:00PM CT (US), February 24th.
We apologize for any inconvenience and appreciate your patience during this improvement process.Please note that you still have telephone and email access to our local offices. We apologize for any inconvenience.
Tambar Group – Professor Product Portal
Professor Product Portal Index
Professor Uttam K. Tambar
The Tambar Group has developed a series of reactions to convert unactivated olefins into value-added products (e.g., allylic amination, allylic alkylation). The transformations developed by Tambar’s lab utilize bis(phenylsulfonyl)sulfur diimide (L511390) as a key component in activating olefins for selective chemical modification.
In addition to developing new chemical reactions, the Tambar Lab synthesizes biologically active natural products and small molecule inhibitors of challenging biological targets for the discovery of novel therapeutic agents.
Tambar group website
For applicable product information, please click below:
For recent articles from the Tambar Laboratory:
Products available at Aldrich from the Tambar Laboratory
Hongli Bao, Uttam K Tambar
Journal of the American Chemical Society 2012-11-14
The enantioselective allylic amination of unactivated terminal olefins represents a direct and attractive strategy for the synthesis of enantioenriched amines...Read More
Hongli Bao, Liela Bayeh, Uttam K Tambar
Angewandte Chemie. International edition in English 2014-02-03
New advances in the functionalization of unactivated olefins with carbon nucleophiles have provided more efficient and practical approaches to convert inexpensive starting materials into valuable products. Recent examples have been reported with stabilized carbon nucleophiles, tethered carbon nucleophiles, diazoesters, and trifluoromethane donors...Read More