Wu Group - Professor Product Portal
Professor Product Portal Index
Professor Xiao-Feng Wu
The Wu Group works with Sigma-Aldrich in providing a portfolio of reagents and systems for carbonylation transformations. Xiao-Feng Wu has recently developed a new CO releasing reagent, TFBen. This reagent has advantages including efficient, non-reacting, and etc. Several synthetic applications with TFBen as CO surrogate have been developed.
Xiao-Feng Wu Group Website
For applicable product information, please click below:
For recent articles from the Xiao-Feng Wu:
Products available at Sigma-Aldrich in collaboration with the Xiao-Feng Wu
Department of Chemistry, Zhejiang Sci-Tech University, Xiasha Campus, Hangzhou 310018, People's Republic of China.
2018 Feb 14;16(7):1065-1067
A DBU-promoted carbonylative cyclization of propargylic alcohols with sulfur was developed. Various 1,3-oxathiolan-2-ones were produced in 61-98% yields under mild conditions in the absence of metal catalysts. TFBen (benzene-1,3,5-triyl triformate) as an efficient and solid CO surrogate and S8 as an ideal sulfur source were emplRead More