Filter Holder Selection Guide
Regardless of the scale or method, membranes must be housed in a device during filtration. Filter housings provide structural support and create a seal around the membrane, preventing filtrate contamination. Reusable housings, (i.e. filter holders) are constructed from either glass, plastic, or metal and must be matched to the diameter of the filter.
Use the selection table below to find the filter holder that fits your needs. Choose based on these criteria:
  • Filter holder material (glass, stainless steel, plastic)
  • Filter disc diameter
  • Pressure-driven filtration or vacuum-driven filtration
Glass
Stainless Steel
Plastic
Filter Diameter Vacuum Vacuum Pressure Vacuum Pressure
13mm   Epifluorescence Filter Holder
Swinny Filter Holder
  Swinnex® Filter Holder, 13 mm
Analytical Stainless Steel Filter Holder Kit, 40 mL funnel, 13 mm
25mm Microanalysis Filter
Holder Kit,
Fritted glass
membrane support
Analytical Stainless Steel Filter Holder Kit, 50 mL funnel, 25 mm
High Pressure Filter Holder
1225 Sampling Manifold
Swinnex® Filter Holder, 25 mm
Solvent Filtering Dispenser
Microanalysis Filter Holder Kit, Stainless steel membrane support
Microsyringe Filter Holder, Luer lock
Microsyringe Filter Holder, NPTF
Filterjet™ Solvent Dispenser
47mm All-Glass Filter Holder Kit, 47 mm, 300mL funnel
Analytical Stainless Steel Filter Holder Kit, 100 mL funnel, 47 mm
Stainless Steel Pressure Filter Holder
Millicup-FLEX Disposable Filtration Kit, Single-Use
Swinnex® Filter Holder, 47 mm
All-Glass Filter Holder Kit, 47 mm, 500mL funnel
Millicup-FLEX Disposable Filtration Kit, Semi-reusable
Classic Glass Filter Holder Kit, Glass frit membrane support, 300 mL funnel
Analytical Stainless Steel Filter Holder Kit, 250 mL funnel, 47 mm
In-line High Pressure Filter Holder
Pressure Vessel, 600 mL
In-line Filter Holder, Polypropylene
Classic Glass Filter Holder Kit, Glass frit membrane support, 500 mL funnel
Classic Glass Filter Holder Kit, PTFE coated membrane support, 300 mL funnel
In-line Filter Holder, Stainless steel
Sterifil® 500 mL Filter Holder
Classic Glass Filter Holder Kit, Stainless steel membrane support, 300 mL funnel
MilliSolve™ Kit, Bottle-to-Bottle Filtration System
Hydrosol™ Filter Holder Kit
90mm All-Glass Filter Holder Kit, 90 mm, Stainless steel membrane support
  Standing Stainless Steel Filter Holder, 90 mm
All-Glass Filter Holder Kit, 90 mm, Glass frit membrane support
142mm     Standing Stainless Steel Filter Holder, 142 mm
Supporting Hardware, Vacuum pumps, and Pressure vessels
Complementing our comprehensive filter holder offering, our supporting hardware, vacuum pumps, and pressure vessels provide robust solutions for a variety of filtration applications and conditions.
Materials