Amicon® Pro Purification System – Antibody Labeling
Label antibodies without losing a single drop — in as fast as one hour
The Amicon® Pro purification system provides the ideal processing tool for small-batch antibody labeling. Antibody cleanup routinely involves the removal of BSA/carrier, labeling, removal of unbound label, dialysis, and final concentration. Using standard dialysis, the procedure can take up to 3 days to complete. 
In contrast, the Amicon® Pro method requires just one hour. Also, thanks to the all-in-one format of the Amicon® Pro device, it eliminates loss due to sample transfers and can significantly improve recovery. 
Buy the Amicon® Pro Purification System
Antibody labeling using Amicon® Pro vs competitor
Click image to enlarge
Generate FITC-labeled antibody in one hour.
Compare the Amicon® Pro Workflow with Traditional Methods
Ab Labeling* Dialysis-based buffer exchange pre/post labeling Centrifugal diafiltration devices - 0.5 mL Amicon® Pro Purification System
Process time 3 days 5 hours 1 hour
Steps required 11 8 5
Sample transfers 3 0 0
Amount Ab required 1 mg 20 μg 20 μg
The antibody labeling protocol consists of four basic steps: antibody cleanup, the labeling reaction, unbound dye removal/buffer exchange, and concentration. The duration of the labeling reaction can vary dependent on individual reagent parameters. The process time presented here is based on biotinylation using the Innolink™ Biotin 354S kit (Catalog. No. 203119)