Nanomaterials for Biomedical Applications
Material Matters 2007, 2.1, 26.
Material Matters 2007, 2.1, 26.
Dendrimers are just one class of nano-sized materials that are explored to develop new life-saving technologies. Single-wall carbon nanotubes are used to make miniaturized sensors for DNA, pathogens, and chemicals – for example glucose sensors. Multi-wall nanotubes, nanoclays, and ceramic nanoparticles are incorporated in ultra-strong polymer nanocomposites that will be used in orthopedic, for example joint-replacement, applications. Nanoparticles can also be used to make nano-textured substrates to support attachment and growth of cells for tissue engineering. Hydroxyapatite nanopowders (677418), incorporated in biodegradable polymer composites1 or deposited on biocompatible substrates2, have been shown to promote adhesion and proliferation of bone-forming (osteoblast) cells. We are looking forward to bring you new nanomaterials tools to accelerate research in biotechnology and medicine – please let us know how we can help: Technical service
Hydroxyapatite Nanopowder (677418)
  • Molecular Formula [Ca5(OH)(PO4)3]x
  • Average Particle Size 150 – 200 nm
  • Surface Area > 9.4 m2/g
  • Available in 5 g pack size
back to top
Materials
References
  1. Kim, S.; Park, M., Jeon, O.; Choi, C.; Kim, B., Biomaterials 2006, 27, 1399.
  2. Sato, M.; Sambito M.; Aslani, A.; Kalkhoran, N.; Slamovich, E.; Webster, T., Biomaterials 2006, 27, 2358.