Attention:
Certain features of sigmaaldrich.com will be down for maintenance Saturday afternoon/evening, February 24th starting at 3:00 pm CT until 9:00 pm CT.
Please note that you still have telephone and email access to our local offices. We apologize for any inconvenience.
Organic Semiconductor Laser Materials
By: Chihaya Adachi, Hajime Nakanotani, Material Matters 2009, 4.3, 74.
Chihaya Adachi*, Hajime Nakanotani
Center for Future Chemistry, Kyushu University, Fukuoka City, Japan
*E-mail: adachi@cstf.kyushu-u.ac.jp
Abstract
Over recent years, a wide variety of challenges aiming for electrical pumping of organic laser diodes have been addressed. However, organic laser diodes have difficulty gaining widespread application due to their high lasing thresholds under electrical pumping. A variety of organic semiconductors have been developed to reduce the lasing threshold, which include not only small molecular materials but also conjugated polymers. In this article, we demonstrate that bis-styrylbenzene derivatives show promising characteristics for very low lasing thresholds and discuss the design considerations for organic lasing molecules.
back to top Back to Top
Introduction
The dissolution of luminescent organic dye molecules in an organic solvent followed by their excitation with high-energy light (greater than their lasing threshold) produces a laser beam. Due to their broad tunabilities, dye lasers have made remarkable advancements.1-5 In recent years, organic solid-state lasers involving the dispersion of an organic dye within a solid/crystal matrix have also been actively studied. In 1989, we proposed an idea to create an organic semiconductor laser diode using triplet excitons of rare earth complexes such as a europium (Eu) complex.6 This system would produce an electrically excited laser with a low threshold by forming a population inversion using the long excitation lifetime of Eu. There have been many studies reporting amplified spontaneous emission (ASE) oscillation from an organic solid thin film and laser oscillation from an organic thin film with an oscillator structure. The thin films in these studies were made by doping a polymer or small-molecule medium with an appropriate laser dye.7-11 These studies provide evidence that organic dyes have suitably high stimulated emission coefficients and that the formation of a solid thin film waveguide can provide the necessary architecture for a solidstate device with a low lasing threshold. More recently, laser oscillations from organic semiconductor dyes, which take electrical excitation into consideration, have also been actively studied.12,13 This interest was generated because organic light-emitting diodes (OLEDs) have achieved an internal luminescence efficiency of nearly 100% through the use of a triplet excited state at the luminescence center. This excited state leads to the realization of an organic semiconductor laser that is an extension of OLEDs a practical study subject of organic electronics.14 In addition, light-emitting transistors have also become capable of producing highly efficient electroluminescence (EL), prompting new electrical excitation developments of organic semiconductors.14-21 In this article, we will discuss materials issues for the production of an organic semiconductor laser. This process requires the development of a new laser material with a focus on a low lasing threshold. Derivatives of bis-styrylbenzene (BSB) have demonstrated extremely good ASE oscillation characteristics in organic solid thin films.15,16 As a result, we will introduce new organic semiconductor materials with similar ASE characteristics in organic solid thin films by using BSB analogs as active materials.
back to top Back to Top
The Mechanism and Goal of Organic LEDs
The external quantum efficiency (ηext) of OLEDs is based on four factors: (i) the ratio of electron and hole injection, transport, and recombination (ν), (ii) exciton formation efficiency (ηr), (iii) internal electroluminescence quantum yield (φp), and (iv) light out-coupling efficiency (ηp) (Figure 1).
Figure 1 OLED luminescence processes (carrier injection, transport, recombination, exciton formation/decay, and light extraction) and their efficiencies. Figures in parentheses represent refractive indices.
Obtaining the highest possible luminescence efficiency requires bringing each of the four factors as close to 100% as possible. Here, ν is a physical quantity containing the ratio of electron and hole injection and transport, as well as the probability of electron and hole recombination. φp can have a value near to 100% by using a material with a high internal luminescence quantum yield. Although ηr is generally low (25% when fluorescent materials are used), the use of phosphorescent materials can theoretically provide an ηr of 75 to 100%. If a device is formed on a common glass substrate, ηp is low at about 20%. Therefore, if fluorescent materials are used as luminescent molecules, ηext is approx 5% at most, but with the use of a triplet exciton as a luminescence transition process can provide a luminescence efficiency theoretically 3 or more times higher. In instances where the intersystem crossing (ISC) probability is about 100%, a luminescence efficiency approximately 4 times higher can be acheived. Figure 2 shows typical luminescent materials used in OLEDs.
Figure 2 Fluorescent and phosphorescent materials used in OLEDs
For Alq3 (Aldrich Prod. No. 444561) and DPVBi, luminescence is found during the transition from the excited singlet state to the ground state. In contrast, for Eu (Aldrich Prod. No. 538957), platinum (Pt) (Aldrich Prod. No. 673625), and iridium (Ir) (Aldrich Prod. No. 688096) complexes, all singlet excitons undergo intersystem crossing into triplet excitons due to the internal heavy atom effect. In the triplet state, a radiative transition occurs with high luminescence efficiency. Use of an Ir compound at the luminescence center reportedly provides a high external EL quantum yield (ηext) of greater than 20%, showing that the internal quantum yield (ηint) reaches almost 100%.22 As described above, the electrons and holes injected into the organic layer result in almost immediate exciton formation.
back to top Back to Top
Laser Active Materials (Fluorescent and Phosphorescent Materials)—Population Inversion Formation at a Low Threshold
To investigate various organic laser materials, we synthesized a variety of styrylamine-, coumarin-, and cyanine-based materials. Here, we will discuss the light amplification characteristics (ASE characteristics) of styrylbenzene derivatives, which we believe to be the most promising laser materials.15,16 Figure 3 presents a selection of the molecules that were studied.
Figure 3 Styrylbenzene-based organic laser materials
Table 1 shows the ASE characteristics of thin films that are 6 wt% doped with styrylbenzene-based luminescent materials under optical excitation with 4,4’-bis[9-dicarbazolyl]-2,2’-biphenyl (CBP) (Aldrich Prod. No. 660124) as the host. Typical BSB which have a dimer skeleton often exhibit especially low threshold (Eth ≅ 1.0 μJ/cm2). Molecules with a heterocycle have an ASE threshold of 20 to 100 μJ/cm2, and molecules with an azomethine skeleton do not cause emission spectrum narrowing even with an excitation energy >100 μJ/cm2. To identify the controlling factors of ASE thresholds, data values for absolute photoluminescence quantum efficiency (φf ), fluorescence lifetime (τf ), radiative decay constant (kr), and triplet-triplet absorption are shown in Table 1.
Table 1 Basic optical physical characteristics of styryl-based laser materials
The ASE thresholds are not directly correlated with φf or τf but are greatly correlated with kr. The magnitude of kr can determine how large the threshold is. 4,4’-bis[(N-carbazole)styryl]biphenyl (BSB-Cz) has extremely good ASE characteristics. The threshold of BSB-Cz is the lowest of the styryl-based fluorescent materials studied so far.15 The luminescence intensity and lifetime do not depend on temperature, so the nonradiative decay is completely inhibited, even at room temperature. The ASE oscillation wavelength of BSB-Cz corresponds to the 0-1 transition in the PL spectrum. The radiative decay constant (kr), stimulated emission cross-section (σem), and absorption crosssection (σABS) are calculated as below. kr (= ηPL/τf ) is calculated with the fluorescence lifetime (τf ) and luminescence quantum efficiency (ηPL) of each codeposited thin film. σem is given by equations (1) and (2) below with the absolute luminescence quantum yield and fluorescence lifetime of each codeposited thin film:24,25
where, Ef (λ) is quantum yield distribution, n (λ) is the refractive index of each wavelength, and τf is fluorescence lifetime. When the refractive index n = 1.8, the absorption cross-section σABS 26 is given by equation (3) below:
where, ε(λ) is the molar absorption coefficient, and NA is Avogadro’s number. kr and σem are strongly correlated with the ASE oscillation threshold Eth in each codeposited thin film. The ASE oscillation thresholds in thin films with the dimmers BSB-Cz, BSB-Me, and BSBOMe as active materials are 1 μJ/cm2 or less. The 6 wt%-BSB-Cz:CBP thin film’s σem is the highest. Although the SBD-4:CBP codeposited thin film is higher in kr and σem than the SBD-1:CBP codeposited thin film, SBD-4:CBP has a high oscillation threshold Eth and the saturation of luminescence intensity is noted at high excitation intensity. This suggests that there is a factor inhibiting luminescence in the 6 wt%-SBD-4:CBP codeposited thin film. Here, the effective stimulated emission cross-section is the difference between the stimulated emission cross-section and loss-related cross-sections (absorption cross-section and excited-state absorption cross-sections) and is given by equation (4) below:
where, σemeff is the effective stimulated emission cross-section, σem is the stimulated emission cross-section, σSS is the singlet excited-state absorption cross-section, and σTT is the triplet excited-state absorption cross-section. The absorption cross-section at the ASE oscillation wavelength of the SBD-4:CBP codeposited thin film is negligible, meaning that the excited-state absorption has a great impact on σemeff. Singlet and triplet excited-state absorption is found in any of the codeposited thin films, and the ASE oscillation wavelength of SBD-1 has triplet excited-state absorption, but not singlet excited-state absorption. In contrast, SBD-4 has singlet and triplet excited-state absorption in the luminescence wavelength range. This result shows that the occurrence of singlet excitation absorption under strong excitation inhibits the formation of population inversion, resulting in no stimulated emission. Figure 4 shows the stimulated emission and absorption cross-sections spectra and excited-state absorption spectrum of the 6 wt%-BSB-Cz:CBP codeposited thin film that has the lowest ASE oscillation threshold. For BSB-Cz, neither the single nor triplet excited-state absorption exists at the ASE oscillation wavelength, providing materials designed to perform at low thresholds.
Figure 4 ASE spectrum and S-S and T-T absorption spectra of BSB-Cz
back to top Back to Top
Development to An Electrically Excitable Device Structure
OLED and FET structures also show promise in realizing an organic semiconductor laser. The OLED structure is basically a double heterostructure to effectively trap electrons and holes as well as trapping generated excitons within the luminescent layer. Advantages of this structure include the ability to use the OLED design guidelines developed so far and the flexibility to select a wide range of organic materials. However, a drawback of this structure is the failure to guide light effectively if a metal electrode is used as the cathode. Therefore, a transparent electrode such as ITO needs to be used at both the anode and cathode. For example, when a transparent device structure with ITO (30 nm)/a-NPD (20 nm)/CBP:BSB [6 wt%] (70 nm)/BCP (20 nm)/Alq3 (20 nm)/MgAg (X nm)/ITO (30 nm)15 is used, a clear ASE oscillation is noted under optical excitation as long as the MgAg layer is between 1 and 3 nm thick (Figure 5).
Figure 5 ASE characteristics under the excitation of light from an electrically excitable laser structure
Recently, even transistor structures have allowed luminescent devices to be constructed. Light emission has actually been observed near the drain electrode by using p-type organic semiconductors such as oligothiophene27, tetracene28 (Aldrich Prod. No. 698415), fluorene-based polymers (Aldrich Prod. No. 571032), PPV-based polymers29 (Aldrich Prod. No. 541443), and thiophene-based oligomers30 (Figure 6).
Figure 6 Organic semiconductor materials for light-emitting transistors
Also, use of a tetraphenylpyrene derivative (TPPy) has dramatically improved luminescence efficiency with an external EL efficiency of 1%.31 These devices are unipolar light-emitting transistors, and their operation mechanism is shown in Figure 7-1. Application of a gate voltage allows holes to accumulate between the electrodes and the organic layer, and an increase in the voltage between the source and drain electrodes allows a region with no hole accumulation (known as a pinch-off region) to be formed near the drain electrode. A strong electric field is created at the interface between the drain electrode and the pinch-off region, accelerating the electron injection.
Figure 7 Schematics and operation mechanisms of light-emitting transistors
In addition, the development of ambipolar organic semiconductors has lead to higher EL efficiency, and some ambipolar devices (Figure 7-2) have been fabricated with polymer organic semiconductors.32,33 Even in a small-molecule system, a clear electroluminescence has been observed (Figure 8) when a single-crystal from a BSB derivative with a simple molecular frame34 is used. Application of an appropriate gate voltage provides electroluminescence between the source and drain electrodes.
Figure 8 Ambipolar FET producing bright electroluminescence
This ambipolar type architecture also has many advantages over OLEDs and shows promise as a future device structure for organic semiconductor lasers. Transistors using organic substances have been studied since the early 1980s when a pioneering organic transistor was first reported.35 As findings about materials and devices have accumulated rapidly, the use of fused polycyclic aromatic compounds such as tetracene and rubrene (Aldrich Prod. No. 554073) have recently achieved an electron field-effect mobility of about 10 cm2/Vs. Electroluminescence from devices using a transistor structure has a high electronic and optical freedom, and are therefore suitable for organic semiconductor lasers, holding promise for future developments.
back to top Back to Top
Materials
References
  1. Sorokin, P. P.; Lankard, J. R. IBM J. Res. Develop. 1966, 10, 162-163.
  2. Schäfer, F. P.; Schmidt, W.; Volze, J. Appl. Phys. Lett. 1966, 9, 306 -309.
  3. Soffer B. H.; McFarland, B. B. Appl. Phys. Lett. 1967, 10, 266-267.
  4. Sorokin, P. P.; Lankard, J. R.; Moruzzi, V. L.; Hammond, E. C. J. Chem. Phys. 1968, 48, 4726-4741.
  5. Sorokin, P. P.; Lankard, J. R.; Moruzzi, V. L.; Lurio, A. Appl. Phys. Lett. 1969, 15, 179-181.
  6. Chihaya Adachi, Student Advanced technical paper, 1989, Nippon Kogyo Shinbun
  7. Tessler, N.; Denton, G. J.; Friend, R. H. Nature 1996, 382, 695-697.
  8. Frolov, S. V.; Ozaki, M.; Gellermann, W.; Vardeny, Z. V.; Yoshino, K. Jpn. J. Appl. Phys. 1996, 35, L1371- L1373.
  9. Hide, F.; diaz-García, M. A.; Schwartz, B. J.; Andersson, M. R.; Pei, Q.; Heeger, A. J. Science 1996, 273, 1833-1836.
  10. Kozlov, V. G.; Bulovic, V.; Burrows, P. E.; Forrest, S. R.; Nature 1997, 389, 362-364.
  11. Berggren, M.; Dodabalapur, A.; Slusher, R. E.; Bao, Z.; Nature 1997, 389, 466-469.
  12. Tessler, N. Adv. Mater. 1999, 5, 363-370.
  13. Yamamoto, H.; Oyamada, T.; Sasabe, H.; Adachi, C. Appl. Phys. Lett. 84, 1401-1403.
  14. Adachi, C.; Baldo, M. A.; Forrest, S. R. J. Appl. Phys. 2001, 90, 5048-8055.
  15. Aimono, T.; Kawamura, Y.; Goushi, K.; Yamamoto, H.; Sasabe, H.; Adachi, C. Appl. Phys. Lett. 2005, 86, 071110-071112.
  16. Nakanotani, H.; Sasabe, H.; Adachi, C. Proc. SPIE Int. Soc. Opt. Eng. 2005, 5937, 59370W-1-7.
  17. Yamamoto, H.; Kasajima, H.; Yokoyama, W.; Sasabe, H.; Adachi, C. Appl. Phys. Lett. 2005, 86, 083502-083504.
  18. Nakanotani, H.; Oyamada, T.; Kawamura, Y.; Sasabe, H.; Adachi, C. Jpn. J. Appl. Phys. 2005, 44, 3659-3662.
  19. Baldo, M. A.; Adachi, C.; Forrest, S. R. Phys. Rev. B 2000, 62, 109671-10977.
  20. Baldo, M. A.; Holmes, R. J.; Forrest, S. R. Phys. Rev. B 2002, 66, 035321.
  21. Nakanotani, N.; Sasabe, H.; Adachi, C. Appl. Phys. Lett. 2005, 86, 213506-1-3.
  22. Adachi, C.; Baldo M. A.; Forrest, S. R. J. Appl. Phys. 2001, 90, 5048-5051.
  23. Yokoyama, W.; Sasabe, H.; Adachi, C. Jpn. J. Appl. Phys. 2003, 42, L1353-1355.
  24. Deshpande, A.; Deshpande, V.; Beidoun, A.; Penzkofer, A.; Wagenblast, G. Chem. Phys. 1990, 142, 123-131.
  25. Holzer, W.; Penzkofer, A.; Schmitt, T.; Hartmann, A.; Bader, C.; Tillmann, H.; Raabe, D.; Stockmann, R.; Hörhold, H.-H. Opt. Quantum Electron 2001, 33, 121-150.
  26. Liu, X.; Py, C.; Tao, Y.; Li, Y.; Ding, J.; Day, M. Appl. Phys. Lett. 2004, 84, 2727-2729.
  27. Shaklee K. L.; Leheny, R. F. Appl. Phys. Lett. 1971, 18, 475-305.
  28. Schön, J. H.; Kloc, Ch.; Haddon R. C.; Batlogg, B. Science 2000, 288, 656-658.
  29. Hepp, A.; Heil, H.; Weise, W.; Ahles, M.; Schmechel, R.; von Seggern, H. Phys. Rev. Lett. 2003, 91, 157406-1-4.
  30. Sakanoue, T.; Fujiwara, E.; Yamada R.; Tada, H. Appl. Phys. Lett. 2004, 84, 3037-3039.
  31. Oyamada, T.; Okuyama, S.; Shimoji, N.; Matsushige, K.; Sasabe, H.; Adachi, C. Appl. Phys. Lett. 2005, 86, 093505.
  32. Oyamada, T.; Uchiuzo, H.; Akiyama, S.; Oku, Y.; Shimoji, N.; Matsushige, K.; Sasabe, H.; Adachi, C. J. Appl. Phys. 2005, 98, 074506-1-7.
  33. Zaumseil, J.; Friend, R. H.; Sirringhaus, H. Nature Materials 2006, 5, 69-74.
  34. Dinelli, F.; Capelli, R.; Loi, M. A.; Murgia, M.; Muccini, M.; Facchetti A.; Marks, T. J. Adv. Mat. 2006, 18, 1416-1420.
  35. Nakanotani, H.; Saito, M.; Nakamura H.; Adachi, C. Appl. Phys. Lett. 2009, 95, 033308.
  36. Kudo, K.; Yamashina, M.; Moriizumi, T. Jpn. J. Appl. Phys. 1984, 23, 130.
Related Links