Attention:
Certain features of sigmaaldrich.com will be down for maintenance Saturday afternoon/evening, February 24th starting at 3:00 pm CT until 9:00 pm CT.
Please note that you still have telephone and email access to our local offices. We apologize for any inconvenience.
Pressure Filter Reactor Systems
Material Matters 2010, 5.2, 55.
Material Matters 2010, 5.2, 55.
Applications
  • Oxidation/reduction reactions where the particulate can be drawn down to a filter cake
  • Nanotechnology where the material must be contained and the filtrate is the actual product or nanoparticle needed
  • Simple Bioreactor as gases can be introduced through pressure head via a pressure or NPT fittings and tubing
  • Paints/pigments other viscous solutions
back to top 
Pressure Filter Reactor Systems
Rugged heavy-wall reactors with a Schott flange with shallow o-ring groove at the top for connection to a jointed head with quick-release clamp. These complete bench top systems include all components required for operation except an IKA® stirrer motor, model RW 20.n (Prod. No. Z645087), which is recommended for all reactor sizes.
back to top 
Choice of Two Models
Pressure Filter Reactor — rated 35 psig @ 100 °C, Ace Threds allow use of vacuum or pressure to drain liquid from reactor, 2 × #15 and 1 × #7 Ace Threds on 60 mm head, 3 × #15 and 1 × #7 Ace Threds on 100 mm head
Jacketed Pressure Filter Reactor — as above with jacket for heating or cooling reaction
For more information, please visit sigma-aldrich.com/labware