Attention:
We are currently upgrading the certain systems that support this website.  You may experience some functionality loss, though we expect the site to be fully functional by 9:00PM CT (US), February 24th.
We apologize for any inconvenience and appreciate your patience during this improvement process.Please note that you still have telephone and email access to our local offices. We apologize for any inconvenience.
Propargyl Claisen Rearrangement
By: Josephine Nakhla, Chemfiles Volume 10 Article 4
Propargyl Claisen Rearrangement
Toste and coworkers reported the use of the gold catalyst [(Ph3PAu)3O]BF4 for the rapid two-step, one-pot sequence of a Claisen rearrangement of a propargyl vinyl ether followed by a reduction of the resulting aldehyde functionality to provide a variety of homoallenic alcohols. The reactions are generally high yielding and the robust catalyst system is able to induce almost complete chirality transfer (in most cases, ee’s were >90%). Low catalyst loadings (1 mol %) and substitution at the alkyne is well tolerated generating the desired allenes in high yields (Scheme 3).
Related Links