Functional, Stereochemically Pure Monomer for ROMP Polymerization
By: Joe Porwoll, Material Matters 2010, 5.1, 1.
Material Matters 2010, 5.1, 1.
Joe Porwoll, President
Aldrich Chemical Co., Inc.
Do you have a compound that you wish Sigma-Aldrich® could list to help materials research? If it is needed to accelerate your research, it matters—please send your suggestion to https://www.sigmaaldrich.com/techservice and we will be happy to give it careful consideration.
Professor Karen Wooley of Texas – A&M University kindly suggested that we offer the exo-5-Norbornene-carboxylic acid (Aldrich Prod. No. 718149) as a product in our catalog. This compound is a functional, stereochemically pure monomer used in ring opening metathesis polymerization (ROMP) synthesis of well-defined polymer materials. The pure exo- monomer shows faster reaction times,1 higher conversion ratios, and better control of ROMP synthesis2 compared to the racemic analog. The carboxylic acid serves as a versatile handle that can be used to prepare a variety of polymers (e.g., glycopolymers) for biomedical applications.3-5
back to top 
Materials
References
  1. Fu, Q.; Seery, T. A. P. Polym. Prepr. 2001, 41 (1), 341-342.
  2. Pollino, J. M.; Stubbs, L. P.; Weck, M. Macromolecules 2003, 36, 2230-2234.
  3. Gordon, E. J.; Gestwicki, J. E.; Strong, L. E.; Kiessling, L. L. Chem. Biol. 2000, 7, 9-16.
  4. Strong, L. E.; Kiessling, L. L. J. Am. Chem. Soc. 1999, 121, 6193-6196.
  5. Owen, R. M.; Gestwicki, J. E.; Young, T.; Kiessling, L. L. Org. Lett. 2002, 4, 2293-2296.