Magnesium Borohydride for Alternative Energy Applications
By: Joe Porwoll, Material Matters 2009, 4.4, 87.
Material Matters 2009, 4.4, 87.
Joe Porwoll
Joe Porwoll, President
Aldrich Chemical Co., Inc.
Prof. Sesha S. Srinivasan of Tuskegee University kindly suggested that we offer Magnesium Borohydride (Aldrich Prod. No. 715247) as a material for alternative energy applications.1 This complex metal hydride, produced by Aldrich Materials Science R&D, is a base-material for advanced energy storage systems with potential for fulfilling the United States Department of Energy's hydrogen storage goals.2 Magnesium borohydride crystallizes in a hexagonal phase that is stable at room temperature. At 185 °C, the material converts to an orthorhombic phase, which persists and is stable upon cooling back to room temperature. Both phases consist of complex networks of cornersharing tetrahedra including a central Mg atom and four BH4 units.3
back to top
  1. Jensen, C. M.; Severa, G.; Ronnebro, E.; Autrey, T.; Orimo, S. Abstracts of Papers, 238th ACS National Meeting, Washington, DC, United States, 2009, FUEL-227.
  2. Read, C.; Thomas, G.; Ordaz, G.; Satyapal, S. Material Matters 2007, 2.2, 3.
  3. Soloveichik, G. Material Matters 2007, 2.2, 11