Attention:
Certain features of sigmaaldrich.com will be down for maintenance Saturday afternoon/evening, February 24th starting at 3:00 pm CT until 9:00 pm CT.
Please note that you still have telephone and email access to our local offices. We apologize for any inconvenience.
CPO-27-Ni Supplemental Information
Explore MOF Applications, Properties and Materials to construct MOFs in the MOF Constructor Tool
Literature suggests that the following materials can be used to prepare CPO-27-Ni. 
Applications
  • Gas adsorption:  Methane gas
Properties
  • Maximum Surface Area: 1060 m2 / g Langmuir Surface Area
  • Pore volume = 0.39 cm3 / g
  • Mean Pore Diameter = 9 Å
Materials
References
  1. Haque, E.; Sung, H. Chemical Engineering Journal 2011, 173(3), 866-872.
  2. Tagliabue, M.; Rizzo, C.; Millini, R.; Dietzel, P.; Blom, R.; Zanardi, S. J. Porous Mater. 2011, 18, 289-296."
Related Links