Attention:
Certain features of sigmaaldrich.com will be down for maintenance Saturday afternoon/evening, February 24th starting at 3:00 pm CT until 9:00 pm CT.
Please note that you still have telephone and email access to our local offices. We apologize for any inconvenience.
Cu2(bptc)(H2O)2 Supplemental Information
Explore MOF Applications, Properties and Materials to construct MOFs in the MOF Constructor Tool
Literature suggests that the following materials can be used to prepare Cu2(bptc)(H2O)2
Applications
  • Gas adsorption:  Hydrogen gas
Properties
  • Maximum Surface Area: 1670 m2 / g BET surface area